• USA Home
  • Z403695 - IKA® Color Squid IKAMAG magnetic plate stirrers

EMAIL THIS PAGE TO A FRIEND
Z403695 Aldrich

IKA® Color Squid IKAMAG magnetic plate stirrers

Synonym: magnetic plate stirrers, magnetic stirrers, stirrer, stirrers

Purchase

Properties

Related Categories Labware, Magnetic Stirrers, Stirrers More...
material   Color Waves top plate design
sterility   non-sterile
packaging   case of 1
  pack of 1
mfr. no.   IKA 2669916
availability   not available in EU
color   none
matrix active group   Non-treated surface treatment

Description

General description

IKA Color Squid IKAMAG

Small magnetic stirrers in various motifs. Recyclable materials with very good chemical resistance provided by glass top and synthetic lower part made of HYTREL.

• Electronically controlled motor
• Speed range: 0-1500 rpm
• Stirring volume: 800 mL (water)

Legal Information

IKA is a registered trademark of IKA-Werke GmbH & Co. KG

IKAMAG is a trademark of IKA-Werke GmbH & Co. KG

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?