• USA Home
  • Z404187 - IKA® MS minishaker microtiter plate attachment

EMAIL THIS PAGE TO A FRIEND
Z404187 Aldrich

IKA® MS minishaker microtiter plate attachment

Purchase

Properties

Related Categories IKA, IKA MS, IKA MS Shakers, IKA Shakers, Labware,
sterility   non-sterile
mfr. no.   IKA L001280
color   none
matrix active group   Non-treated surface treatment

Description

Legal Information

IKA is a registered trademark of IKA-Werke GmbH & Co. KG

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?