• USA Home
  • Z404772 - IKA® RW 11 "Lab egg" stirrer base plate

EMAIL THIS PAGE TO A FRIEND
Z404772 Aldrich

IKA® RW 11 "Lab egg" stirrer base plate

small base plate, 1/cs

Purchase

Properties

Related Categories IKA, IKA Lab Egg Stirrers, IKA Stirrers, Labware, Manufacturer Browser,
sterility   non-sterile
packaging   case of 1
  pack of 1
mfr. no.   IKA 29725.00
rod L × diam.   360 mm × 10 mm
color   none
matrix active group   Non-treated surface treatment
suitability   suitable for (RW 11 "Lab egg" stirrer)

Description

Legal Information

IKA is a registered trademark of IKA-Werke GmbH & Co. KG

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?