• USA Home
  • Z404950 - Organic Reactions in Aqueous Media
EMAIL THIS PAGE TO A FRIEND
Z404950 Aldrich
Organic Reactions in Aqueous Media
  •  ISBN-10 0471163953
  •  ISBN-13 978-047116395-4
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   C. Li, John Wiley & Sons, Ltd, 1997, hard cover
mfr. no.   Wiley
Description
General description
Rather than using hazardous substances, chemists are now looking to using water as a solvent in synthesis. This book provides readers with a general understanding of the current thinking in the field, from basic theory to synthetic application. It presents information necessary for designing efficient synthesis using water.
Table of Contents
Pericyclic Reactions.
Nucleophilic Additions and Substitutions.
Metal- Mediated Reactions.
Transition-Metal-Catalyzed Reactions.
Oxidations and Reductions.
Industrial Applications.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?