• USA Home
  • Z404969 - Reductions by the Alumino- and Borohydrides in Organic Synthesis, 2nd ed.
EMAIL THIS PAGE TO A FRIEND
Z404969 Aldrich
Reductions by the Alumino- and Borohydrides in Organic Synthesis, 2nd ed.
  •  ISBN-10 0471190365
  •  ISBN-13 978-047119036-3
Purchase
Properties
Related Categories Books and Software, Inorganic and Organometallic Books, Labware More...
publication info   J. Seyden-Penne, H.C. Brown (foreword), John Wiley & Sons, Ltd, 1997, 224 pp., hard cover
mfr. no.   Wiley
Description
General description
This updated second edition is a guide to the selection of reducing reagents in organic synthesis. It is an indispensable working resource for organic synthetic chemists and the only reference focusing exclusively on aluminohydrides and borohydrides and their derivatives.
Table of Contents
Description and Characteristics of the Main Reagents.
Cleavage of the Carbon-Heteroatom Single Bond.
Reduction of Double Bonds.
Reduction of Triple Bonds.
Other Derivatives.
Synoptic Tables.
References.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?