• USA Home
  • Z405418 - Radicals, Ion Radicals, and Triplets: The Spin-Bearing Intermediates of Organic Chemistry
EMAIL THIS PAGE TO A FRIEND
Z405418 Aldrich
Radicals, Ion Radicals, and Triplets: The Spin-Bearing Intermediates of Organic Chemistry
  •  ISBN-10 0471190357
  •  ISBN-13 978-047119035-6
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   N.L. Bauld, John Wiley & Sons, Ltd, 1997, 240 pp., hard cover
mfr. no.   Wiley
Description
General description
This overview of all major types of spin-bearing radicals was written by an internationally acknowledged expert in the field with over two decades′ research experience with radicals--important intermediates in organic reactions. While many new synthetic methods involving radicals have been developed and discussed in the past five years, this is the only book focusing on the physical-organic aspects of radical chemistry.
Table of Contents
Basic Concepts of Free Radicals.
Radical Reactions.
The Characterization of Radicals and Radical Pairs by ESR and CIDNP.
Anion Radicals.
Cation Radicals.
Ion Radical Pairs and Electron Transfer.
Triplets and Higher Multiplets.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?