• USA Home
  • Z405868 - Matheson general-purpose gas regulator
EMAIL THIS PAGE TO A FRIEND
Z405868 Aldrich
Matheson general-purpose gas regulator
single-stage, CGA Inlet size 590, 5-125 psig delivery pressure
Purchase
Properties
Related Categories Gas Equipment Product Guide, Gas Regulators, General-purpose gas regulator, Labware, Manufacturer Browser,
mfr. no.   Matheson Model 18
measuring range   5-125 psig delivery pressure
CGA Inlet size   590
Description
General description
Economical regulators for use with non-corrosive gases. U.L. listed. External relief valve protects regulator from overpressure.
Regulator selection:
Single-stage
regulators reduce source pressure in a single step and are suited for applications where minor fluctuations in delivery pressure is acceptable, such as manufacturing and purging operations.
Dual-stage regulators reduce source pressure in two steps and are suited for applications that require constant pressure, such as instrumentation.
Specifications:
• Forged brass body/chrome-plated brass bonnet
• Nylon reinforced neoprene diaphragm
• PTFE seats/neoprene seals
• 3,000 psig max. inlet pressure
• Temp. range: -20 to 140 °F
• Outlet needle valve with 1/4 in. NPTM connection
Legal Information
Matheson is a trademark of Matheson Tri-Gas, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
29568-U CGA-590 Gas Cylinder Pressure Regulator, 1-stage high purity, parameter 0-100 psi
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?