• USA Home
  • Z406023 - Matheson Monel® corrosive gas regulator with cross-purge assembly
EMAIL THIS PAGE TO A FRIEND
Z406023 Aldrich
Matheson Monel® corrosive gas regulator with cross-purge assembly
CGA Inlet size 660, 3-200 psig delivery pressure
Purchase
Properties
mfr. no.   Matheson 3220-660/4775
measuring range   3-200 psig delivery pressure
CGA Inlet size   660
Description
General description
For use with Cl2 and oxidizers.
The combination of Monel regulator components and proper purging with the cross-purge assembly makes these units virtually impervious to attack from strong acid forming gases that include hydrogen chloride, chlorine, hydrogen fluoride, hydrogen bromide and silicon tetrafluoride. Cross-purge assembly eliminates the escape of toxic and flammable gases when changing cylinders and permits purging system with dry, inert gas to prevent corrosion due to gas hydrolysis.
Specifications:
• Single-stage
• Monel body, diaphragm, and gauges
• KEL-F® seats
• 3,000 psig max. inlet pressure
• Temp. range: -20 to150 °F
• Outlet Monel needle valve with 1/4 in. NPTM connection
Request Technical Information Bulletin No. AL-202 for cross-purge techniques.
Legal Information
Matheson is a trademark of Matheson Tri-Gas, Inc.
Monel is a registered trademark of Huntington Alloys Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
Z562580 Aldrich® corrosive gas regulator with electroless nickel-plated body and cross purge assembly, inlet CGA 660
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?