• USA Home
  • Z406090 - Digi-Block® Jr. digital block heater
EMAIL THIS PAGE TO A FRIEND
Z406090 Aldrich
Digi-Block® Jr. digital block heater
AC input 120 V
Purchase
Properties
Related Categories Barnstead Thermolyne, Barnstead Thermolyne Block Heaters, Barnstead Thermolyne Heating Equipment, Block Heaters, Digi-Block block heaters,
feature   capacity 1 block
  temp. range≤ 110 °C by 0.1 °C
mfr. no.   Barnstead International 5111
H × L × W   15 cm × 16 cm × 11 cm
output   100 W
Description
General description
Compact design saves bench space. Provides safe, dry heating for clinical and general chemistry applications. Interchangeable heating blocks accommodate test tubes, microcentrifuge tubes, and cuvettes. Order blocks separately.
• Control panel displays "actual" and "set" temperatures
• Non-volatile memory retains temperature settings
• Aluminum and SS housing will not melt or rust
• Massive aluminum heating plate assures even heating
• Platinum RTD probe fits into heating blocks next to samples for precise temperature control
• Auto-off timer, 0-99.9h
• "Block Hot" warning light
Legal Information
Digi-Block is a registered trademark of Laboratory Devices, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?