• USA Home
  • Z406406 - Basic Gas Chromatography: Techniques in Analytical Chemistry
EMAIL THIS PAGE TO A FRIEND
Z406406 Aldrich
Basic Gas Chromatography: Techniques in Analytical Chemistry
  •  ISBN-10 0471172618
  •  ISBN-13 978-047117261-1
Purchase
Description
General description
Offers thorough coverage of basic principles and techniques of modern gas chromotography, serving as a primer/working reference for bench chemists and as a text for upper-level undergraduate and graduate students. Theoretical issues are explained without complicated equations and derivations and always in terms of how they relate to practical operating principles. Coverage includes both capillary column and packed column chromotography, as well as cutting-edge topics such as chiral analysis and solid phase microextraction. Includes step-by-step guidelines and trouble shooting tips, reference appendices, and use of new IUPAC terms cross-referenced to traditional terms and symbols. From Book News, Inc., Portland, OR
Table of Contents
Instrument Overview.
Basic Concepts and Terms.
Stationary Phases.
Packed Columns and Inlets.
Capillary Columns and Inlets.
Detectors.
Qualitative and Quantitative Analysis.
Programmed Temperature.
Special Topics.
Troubleshooting GC Systems.
Appendices.
Indexes.
Other Notes
Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?