• USA Home
  • Z406716 - Aldrich® CH109 Chemical Calculator
EMAIL THIS PAGE TO A FRIEND
Z406716 Aldrich
Aldrich® CH109 Chemical Calculator
on 31/2 in. diskette
Purchase
Description
General description
This program functions not only as an advanced scientific calculator, but also calculates molecular weights, % C, % H, % isotope, and many other calculations. It handles complex expressions, chemical and biochemical abbreviations, and offers users the benefit of speed and accuracy in chemical, peptide, and isotope calculations.
System Requirements
Windows95, NT 3.51
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
CH109 is a trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?