• USA Home
  • Z407844 - Safetypette pipette filler
EMAIL THIS PAGE TO A FRIEND
Z407844 Aldrich
Safetypette pipette filler
pkg of 1 ea
Purchase
Properties
Related Categories Labware, Liquid Handling Equipment, Pipette Controllers and Pumps More...
material   silicone bulb (squeeze)
packaging   pkg of 1 ea
volume   1-100 mL
Description
General description
• Superior control for 1 to 100 mL pipettes for both left or right handed users
• Silicone squeeze bulb and thumb lever controls both aspirate and dispense modes
• Autoclavable
Supplied with a 0.45 μm membrane filter and pipette cones for safety color-coding in white, mint, mauve, and pink.
Legal Information
Safetypette is a trademark of Jencons Scientific, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?