• USA Home
  • Z526363 - Snap-on® five drawer storage cabinet
EMAIL THIS PAGE TO A FRIEND
Z526363 Aldrich
Snap-on® five drawer storage cabinet
dimensions: 15 W × 14 D × 16 in. H.
Purchase
Properties
Related Categories Boxes, Containers, Labware, Manufacturer Browser, Snap-on,
mfr. no.   Snap-On KRP5CAB
Description
General description
Black, heavy-duty PP cabinet with five removable drawers. Each drawer features sixteen translucent section dividers for unlimited customizing options. Corrosion-proof and resistant to common solvents. Units can be stacked or wall mounted.
Cabinet dim.: 15 W × 14 D × 16 in. H.
Legal Information
Snap-on is a registered trademark of Snap-on, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?