• USA Home
  • Z527351 - Matheson high-purity gas regulator
EMAIL THIS PAGE TO A FRIEND
Z527351 Aldrich
Matheson high-purity gas regulator
single-stage, CGA Inlet size 540, 4-100 psig delivery pressure
Purchase
Properties
Related Categories Gas Equipment Product Guide, Gas Regulators, High-purity gas regulator, Labware, Manufacturer Browser,
mfr. no.   Matheson 3539-540
measuring range   4-100 psig delivery pressure
CGA Inlet size   540
Description
General description
For use with non-corrosive gases in applications where maintenance of system purity is critical. Bonnet ported and threaded to pipe gases away from work area.
Regulator selection:
Single-stage
regulators reduce source pressure in a single step and are suited for applications where minor fluctuations in delivery pressure is acceptable, such as high-purity sampling, manufacturing, and purging operations.
Dual-stage regulators reduce source pressure in two steps and are suited for applications that require constant pressure, such as GC instrumentation using PID, HID, or ECD detection methods.
Specifications:
• Nickel-plated brass barstock body and bonnet
• 316 stainless steel diaphragms
• PFA seat (1st stage) TFE seat (2nd stage)/metal to metal seals
• 3,000 psig max. inlet pressure
• Temp. range: -40-165°F
• Outlet diaphragm valve with 1/4 in. compression connection
Legal Information
Matheson is a trademark of Matheson Tri-Gas, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?