• USA Home
  • Z527556 - Matheson stainless steel gas regulator
EMAIL THIS PAGE TO A FRIEND
Z527556 Aldrich
Matheson stainless steel gas regulator
dual-stage, CGA Inlet size 660, 4-100 psig delivery pressure
Purchase
Properties
Related Categories Gas Equipment Product Guide, Gas Regulators, Labware, Manufacturer Browser, Matheson,
mfr. no.   Matheson 3810
measuring range   4-100 psig delivery pressure
CGA Inlet size   660
Description
General description
For use with non-corrosive and semi-corrosive gases and in high-purity control applications. Bonnet ported and threaded to pipe gases away from work area.
Regulator selection:
Single-stage
regulators reduce source pressure in a single step and are suited for applications where minor fluctuations in delivery pressure is acceptable, such as high-purity sampling, manufacturing, trace analysis, and purging operations.
Dual-stage regulators reduce source pressure in two steps and are suited for applications that require constant pressure, such as GC instrumentation using PID, HID, or ECD detection methods, calibration standards, semiconductor applications, and doping operations.
Specifications:
• 316 stainless steel construction
• Nickel-plated brass bonnet
• PFA seats, metal to metal seals
• 3,000 psig max. inlet pressure
• Temp. range: -40-165 °F
• Outlet diaphragm valve with integral Gyrolok® 1/4 in. compression connection
Legal Information
Gyrolok is a registered trademark of Hoke Inc.
Matheson is a trademark of Matheson Tri-Gas, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?