| Related Categories | Accelerator Free Disposable Nitrile Gloves, Best Gloves, Best N-DEX Free Disposable Nitrile Gloves, Best N-DEX Free disposable nitrile gloves, Disposable Gloves, |
| size | S |
| mfr. no. | Best Manufacturing 7705PFTS-07 |
| packaging | pkg of 100 ea |
| L × thickness | 9.5 in. × 4 mil |
| color | green |
Developed by Best to the same exacting standards of its N-DEX family of hand protection.
N-DEX Free′s accelerator-free formulation creates a luminous, fun, yet PPE-conscious, high-visibility green glove.
• Powder-free, ambidextrous
• Free from accelerators common in all disposable nitrile gloves, which have been linked to workplace conditions like Type IV dermatitis
• Free from natural rubber latex and Type I allergic reactions linked to latex proteins
Best is a registered trademark of Showa Glove Co.
N-DEX is a registered trademark of Showa Glove Co.