• USA Home
  • Z563544 - Büchi® heating bath for R-210/215 evaporators
EMAIL THIS PAGE TO A FRIEND
Z563544 Aldrich
Büchi® heating bath for R-210/215 evaporators
AC input 100 - 120 V
Purchase
Properties
Related Categories Buchi, Buchi Baths, Buchi Parts List, Buchi Rotary Evaporator Accessories, Evaporators,
description   Model B-491
feature   CSA approved
mfr. no.   Büchi, 048201
availability   not available in EU
parameter   20-180 °C temp. limit
capacity   ≤4 L
Description
General description
The heating bath, for water or oil covers a wide temperature range. A continuous display shows the actual temperature and the temperature setpoint. It has a cordless power supply so it is not necessary to unplug connections when filling or emptying the bath.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?