• USA Home
  • Z566985 - IKA® RV 10 rotary evaporator, Digital
EMAIL THIS PAGE TO A FRIEND
Z566985 Aldrich
IKA® RV 10 rotary evaporator, Digital
diagonal condenser, plastic-coated glass, AC input 115 V, pack of 1
Synonym: rotary evaporator condensers, rotovap condensers
Purchase
Properties
Related Categories Evaporators, IKA Rotary Evaporators, Labware More...
material   plastic-coated glass
packaging   pack of 1
parameter   20-280 rpm speed
W × H × D   500 mm × 430 mm × 410 mm
bath capacity   3 L
weight   20 kg
Description
General description
The IKA RV 10 rotary evaporators have integrated heating baths and an ergonomic, space-saving design. Each version features a motorized lifting mechanism that raises the evaporator piston out of the heating bath in case of power outage.
Other safety features also include interval operation, smooth start, end position recognition for the evaporator piston, and a timer function.
The RV 10 evaporators are fitted with a water/oil bath that includes a powerful 1300 Watt heater capable of delivering temperatures from room temperature to 180°C.
Digital model:
• The digital version has accurate digital and temperature display
• Temperature control of heating bath by micro controller
• Drive capable of right-left interval operation
• Infrared interface for data transfer from heating bath to drive unit
• Precision controlled via the RS 232 interface for remote PC operation
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Content
IKA Evaporators [VIDEO]
From our library of Related Content, Sigma-Aldrich presents IKA Evaporators [VIDEO]
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?