• USA Home
  • Z567027 - IKA® RV 10 rotary evaporator, Control

EMAIL THIS PAGE TO A FRIEND
Z567027 Aldrich

IKA® RV 10 rotary evaporator, Control

diagonal condenser, plastic-coated glass, 1/cs, plastic-coated glass, AC input 115 V, pack of 1

Synonym: rotary evaporator condensers, rotovap condensers

Purchase

Properties

Related Categories Evaporators, IKA Rotary Evaporators, Labware More...
material   plastic-coated glass
packaging   pack of 1
limit   20-280 rpm speed
W × H × D   500 mm × 430 mm × 410 mm
bath capacity   3 L
weight   21.5 kg

Description

General description

The IKA RV 10 rotary evaporators have integrated heating baths and an ergonomic, space-saving design. Each version features a motorized lifting mechanism that raises the evaporator piston out of the heating bath in case of power outage. Other safety features also include interval operation, smooth start, end position recognition for the evaporator piston, and a timer function.

The RV 10 evaporators are fitted with a water/oil bath that includes a powerful 1300 Watt heater capable of delivering temperatures from room temperature to 180 °C.

Control model
• The control version has a digital display showing target and actual temperatures.
• Solvent-resistant PTFE seals
• Supplied complete with a Woulff bottle to protect the vacuum pump against solvent condensation.
• Integrated vacuum controller, USB interface and graphical display
• Precision controlled via the RS 232 interface for remote PC operation
• Color graphic display
• Automatic ventilation and integrated cooling water monitoring
• Distillation parameters programmable and stored for standard distillations

Legal Information

IKA is a registered trademark of IKA-Werke GmbH & Co. KG

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

IKA Evaporators [VIDEO]

From our library of Related Content, Sigma-Aldrich presents IKA Evaporators [VIDEO]

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?