• USA Home
  • Z567795 - Pressure monitor set for 35 mL Q-Tube
EMAIL THIS PAGE TO A FRIEND
Z567795 Aldrich
Pressure monitor set for 35 mL Q-Tube
1/4 in. NPT port
Synonym: humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
Purchase
Properties
Related Categories Labware, Pressure Vessels, Q-Tube Pressure Tube Reactors More...
mfr. no.   Q Labtech, 100-PRMSET-N35SS
Description
General description
Designed for pressure monitoring and gas filling (e.g. hydrogenation).
The set contains all items needed for immediate use, including one aluminum adapter, one pressure gauge (300 psi), one glass pressure tube-35 mL, ten septa.
Legal Information
Q-Tube is a trademark of Q Labtech LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?