• USA Home
  • Z602124 - Horst Pt 100 temperature sensors
EMAIL THIS PAGE TO A FRIEND
Z602124 Aldrich
Horst Pt 100 temperature sensors
parameter ≤250 °C temperature
Purchase
Properties
Related Categories Heating, Heating Mantles, Horst, Horst Heating Mantles, Labware,
feature   CE compliant
  with 1 M PTFE-insulated interconnection feeder and plug
mfr. no.   Horst
parameter   ≤250 °C temperature
sensor diam. × L   3 mm × 30 mm
Description
General description
For use with Horst temperature regulators.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?