• USA Home
  • Z603279 - Normag solvent recirculation still
EMAIL THIS PAGE TO A FRIEND
Z603279 Aldrich
Normag solvent recirculation still
volume 500 mL
Purchase
Description
General description
Evaporating Flask with conical joint cone NS 29/32, receiving flask with joint socket NS 14/23. PFTE double spindle valve, completed with lateral adapter with Rotulex connection free of grease. Spherical ground joint clamp, Stainless steel spiral heat exchanger. Metal thread GL 14 and 2 threaded GL 14 hose connectors for cooling water.
Legal Information
Normag is a trademark of Normag Labor- und Prozesstechnik GmbH
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?