• USA Home
  • Z639915 - KNF Laboxact vacuum systems -SEM series
EMAIL THIS PAGE TO A FRIEND
Z639915 Aldrich
KNF Laboxact vacuum systems -SEM series
KNF SEM 842
Purchase
Properties
Related Categories Diaphragm Vacuum Pumps, KNF, Laboxact vacuum systems -SEM series, Labware, Manufacturer Browser,
AC input   220 V
feature   CE compliant
  <2 mbar final vacuum
mfr. no.   KNF SEM 842
availability   not available in USA and Canada
parameter   34 L/min pump flow
Description
General description
The pump uses chemical resistant membranes and the systems are ideally suitable for use with rotary evaportors.
Complete vacuum system consisting of membrane pump unit, high performance condenser, manual control valve, manual vacuum release valve and digital vacuum meter mounted onto a base plate.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?