• USA Home
  • Z645001 - IKA® MS 3 basic shakers

EMAIL THIS PAGE TO A FRIEND
Z645001 Aldrich

IKA® MS 3 basic shakers

universal small shaker, 115 V

Synonym: lab shakers, laboratory shakers, microplate shaker, orbital shaker, vortexer

Purchase

Properties

Related Categories IKA, IKA MS Shakers, IKA Shakers, Labware, Manufacturer Browser,
AC input   115 V, US 2-pin plug
feature   CSA approved
packaging   pkg of 1 ea
mfr. no.   IKA 3617001
availability   not available in EU
W × H × D   148 mm × 63 mm × 205 mm
weight   2.9 kg

Description

General description

IKA® MS 3 Basic Shaker

Compact, universal small shaker suitable for shaking tasks with all small vessels and microtiter plates.

• Wide range of attachments
• Attachment detection
• Continuous or touch operation (with standard attachment)
• Stable in all speed ranges
• Sturdy zinc die cast casing

• Two operating modes:

Mode A (safe mode with attachment detection) the maximum speed of 3000 rpm is only reached with the standard attachment in touch mode. When using other attachments the speed is limited to 1,300 rpm.

Mode B (without attachment detection) a speed of 3000 rpm is possible with all attachments.

Kit contents:shaker MS 3 basic, power supply, standard attachment MS 3.1, universal attachment MS 3.3, one-hand insert MS 1 & operating instructions

Legal Information

IKA is a registered trademark of IKA-Werke GmbH & Co. KG

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

accessory

Product #

Description

Add to Cart

Z676098 IKA® MS 3 minishaker microtiter plate attachment, MS 3.4 microtiter attachment, 1/cs
Z676101 IKA® MS 3 minishaker standard attachment, MS 3.1 standard attachment, 1/cs
Z676128 IKA® MS 3 minishaker universal attachment, MS 3.3 universal attachment, 1/cs

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?