• USA Home
  • Z645648 - Microflex® 93-833 UltraSense nitrile gloves

EMAIL THIS PAGE TO A FRIEND
Z645648 Aldrich

Microflex® 93-833 UltraSense nitrile gloves

size M

Purchase

Properties

Related Categories 93-833 UltraSense nitrile gloves, Disposable Gloves, Disposable Nitrile Gloves, Gloves, Labware,
material   blue gloves
  blue nitrile
size   M
sterility   non-sterile
feature   CE compliant
  disposable
  latex free: no
  powder-free
packaging   package of 100 ea
mfr. no.   MICROFLEX, 93-833-080
L × thickness   9.7 in. (245 mm) × 3.5 mil (0.09 mm)

Description

Other Notes

Microflex UltraSense is in the process of re-branding. The new product name will become Microflex 93-833. Changes effect name and packaging only. No change in material specifications.

General description

A soft nitrile glove with a latex fit! UltraSense has a precise fit, with ultra comfort and unwavering grip, conforming to your hand like latex. Unmatched quality and comfort in an affordable nitrile glove. UltraSense forms to your hand like latex, without the associated risks, and has elasticty unlike any other nitrile.

High quality nitrile UltraSense provides reliable barrier protection against many hazardous and infectious substances.

• Proprietary, ultra-soft formulation provides a comfortable fit and feel
• Forms to your hand to allow full range of motion
• Unique finishing process delivers an outstanding grip for both small utensils and large instruments in wet and dry environments
• Eliminates the possibility of natural rubber latex allergic reactions

Thickness:
• Cuff: 0.07 mm (2.7 mil)
• Palm: 0.09 mm (3.5 mil)
• Finger: 0.12 mm (4.7 mil)
• Length: 245 mm (9.7")

Legal Information

Microflex is a registered trademark of CIT Healthcare LLC

UltraSense is a trademark of Microflex Medical Corp.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?