• USA Home
  • Z677078 - Millex-GN syringe filter units, disposable with Luer fitting
EMAIL THIS PAGE TO A FRIEND
Z677078 Aldrich
Millex-GN syringe filter units, disposable with Luer fitting
pore size 0.45 μm, diam. 33 mm, non-sterile
Purchase
Properties
Related Categories Filtration Supplies, Labware, Manufacturer Browser, Millex-GN syringe filter units, disposable with Luer fitting, Millipore,
material   Nylon membrane
  polypropylene housing
sterility   non-sterile
packaging   pack of 250
mfr. no.   Millipore SLHN033NB
  Millipore, SLHN033NB
diam.   33 mm
volume   ≤100 mL
pore size   0.45 μm
fitting   inlet female Luer (lock)
  outlet male Luer
Description
Application
Recommended for HPLC solvents and solutions; not recommended for protein solutions because of high non-specific binding.
General description
Units contain filter membranes in plastic housings with Luer lock inlet and Luer slip outlet fittings. Hold-up volume after air purge is 0.80 mL.
Suitability
Suitable for clarifying aqueous or organic solutions.
Price and Availability
Pharma QC
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?