• USA Home
  • Z677892 - Amicon Ultra 0.5 mL centrifugal filters
EMAIL THIS PAGE TO A FRIEND
Z677892 Aldrich
Amicon Ultra 0.5 mL centrifugal filters
MWCO 30 kDa
Purchase
Properties
Related Categories Amicon Ultra Centrifugal Filter Units, Centrifuge Filtration, Filtration Supplies, Labware
material   polypropylene tube
  regenerated cellulose membrane
  styrene-butadiene housing
mfr. no.   Millipore, UFC503024
pore size   30 kDa MWCO
Description
Frequently Asked Questions
Frequently Asked Questions are available for this Product.
General description
With their vertical membrane design, Amicon Ultra 0.5 mL filters deliver great performance and lower spin times.
• Concentrates 500 μL down to 15 μL
• Concentration factor 25 x to 30 x
• High sample recovery (greater than 90% of dilute starting solution)
• Fast processing time - 10 to 30 minutes
• Consistent recoveries thanks to the reverse spin capability
• Reverse spin capability provides consistent recoveries
The Amicon Ultra-0.5 device is supplied with two microcentrifuge tubes. During operation, one tube is used to collect filtrate; the other tube is to recover the concentrated sample.
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?