• USA Home
  • Z684465 - BRAND® Dispensette® Organic bottle-top dispenser, analog-adjustable volume

EMAIL THIS PAGE TO A FRIEND
Z684465 Aldrich

BRAND® Dispensette® Organic bottle-top dispenser, analog-adjustable volume

10-100 mL, without SafetyPrime

Synonym: bottle top dispensers

Purchase

Properties

Related Categories BRAND, BRAND Dispensers, BRAND Dispensette Organic Bottle-Top Dispenser, BRAND Dispensette bottle-top dispenser, Dispensers,
feature   DE-M marking
  without SafetyPrime
mfr. no.   BRAND, 4730170 EU
  BrandTech, 4731170 US
measuring range   10-100 mL

Description

General description

Dispensette Organic (yellow color-code): for dispensing organic solvents including: Chlorinated and fluorinated hydrocarbons (e.g., trichlorotrifluoroethane and dichloromethane), concentrated acids (e.g., HCI and HNO3), trifluoroacetic acid (TFE), tetrahydrofuran (THF) and peroxides.

• Easy to dismantle for cleaning, with replaceable filling valves
• Autoclavable at 121°C
• Fast volume adjustment with analog slide
• Simple calibration adjustment with supplied tool
• SafetyPrime recirculation valve reduces reagent waste and the safety discharge system reduces the risk of inadvertent dispensing and splashes
• To allow the bottle label to face the user, the valve block can be rotated 360°
• A telescopic filling tube adjusts easily to different size bottles

The parts of the Dispensette Organic which are in contact with medium are made of: Borosilicate glass, ceramic, tantalum, ETFE, FEP & PFA. The discharge tube safety screw cap is PP.

Operating limits:
• Vapor pressure max. 500 mbar
• Viscosity max. 500 mm2/s
• Temperature max. 40°C
• Density max. 2.2 g/cm3
• Accuracy: ±0.5%, precision ±0.1%

Linkage

See the Compatibility Chart to choose the correct bottle-top dispenser.

Other Notes

BR4730170 (India)

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Dispensette is a registered trademark of BRAND GMBH + CO KG

SafetyPrime is a trademark of BRAND GMBH + CO KG

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?