• USA Home
  • Z684643 - Ace stirrer bearing, mechanical seal
EMAIL THIS PAGE TO A FRIEND
Z684643 Aldrich
Ace stirrer bearing, mechanical seal
for 19 mm shaft, male Joint: ST/NS 45/50
Purchase
Properties
Related Categories Glass Bearing Adapter, Glass Stirring Equipment, Glassware, Labware, Stirrer bearing, mechanical seal More...
material   PTFE
Joint   male Joint: ST/NS45/50
mfr. no.   Ace Glass, 8050-06
Description
General description
Newly designed mechanical stirrer bearing for overhead stir motors. Non-shedding bearing is manufactured from PTFE, Rulon® and PEEK materials.
Equally good in both vacuum and pressure applications as low as -0.6 Torr. Rated for up to 400 rpm with both glass (polished rather than precision ground is best) and stainless steel shafts of 10, 19 and 28 mm diameter.
Legal Information
Rulon is a registered trademark of Saint-Gobain Performance Plastics Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?