• USA Home
  • Z684716 - Ace PTFE filter reactor
EMAIL THIS PAGE TO A FRIEND
Z684716 Aldrich
Ace PTFE filter reactor
capacity 500 mL, head Joints: ST/NS 24/40 (4), #50 valved bottom filter assembly
Purchase
Properties
Related Categories Flasks, Labware, Reaction Flasks More...
material   PTFE
  flask
Joint   Joints: ST/NS head24/40 (4)
sterility   non-sterile
feature   cap: no
packaging   case of 1
mfr. no.   Ace Glass, 6390-05
capacity   500 mL
Description
General description
All PTFE filter reactor; an extension of the famous Ace borosilicate glass filter reactor product line. The head, head joints, body, stirrer bearing, valved bottom filter assembly, and stir shaft with agitator are all PTFE. Filter support is polypropylene.
Large, Ace-Thred bottom filter assembly facilitates easy filtrate recovery & cleaning. The 1,000 mL reactor includes a Halar- coated coil for heating or cooling. These reactors are designed for fluorine chemistry work as well as any work using strong alkalis where glass cannot be used.
Legal Information
Ace-Thred is a trademark of Ace Glass, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?