• USA Home
  • Z684759 - Ace spherical reaction flask with three vertical necks
EMAIL THIS PAGE TO A FRIEND
Z684759 Aldrich
Ace spherical reaction flask with three vertical necks
capacity 3 L
Purchase
Properties
Related Categories Flasks, Labware, Reaction Flasks More...
material   borosilicate glass
  three-neck (Vertical)
Joint   Joints ST/NS24/40 (3)
description   0-10 mm zero dead space bottom valve
sterility   non-sterile
feature   cap: no
packaging   case of 1
mfr. no.   Ace Glass, 6483-120
capacity   3 L
Description
General description
Heavy wall, spherical reaction flask with three vertical necks. Flask has a Zero Dead Space (ZDS) bottom valve. The valve features no material hang-up in the fully open position. When closed, the valve seats flush with the bottom of the flask.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?