• USA Home
  • Z684899 - Ace spherical reaction flask
EMAIL THIS PAGE TO A FRIEND
Z684899 Aldrich
Ace spherical reaction flask
capacity 3 L, 100 mm diam. O-ring grooved flange, valve size 0-20 mm , zero dead space
Purchase
Properties
Related Categories Flasks, Labware, Reaction Flasks More...
material   borosilicate glass
sterility   non-sterile
packaging   case of 1
mfr. no.   Ace Glass, 6540-106
capacity   3 L
valve size   0-20 mm , zero dead space
Description
General description
Heavy wall, spherical reaction flask with Schott grooved flange opening. Flask has a Zero Dead Space (ZDS) bottom valve. The valve features no material hang-up in the fully open position. When closed, the valve seats flush with the bottom of the flask. Includes one CAPFE O-ring.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?