EMAIL THIS PAGE TO A FRIEND
Z684961 Aldrich
Ace mixing baffle
shaft O.D. × L 10 mm × 330 mm, fits #11 Ace-Thred adapter, PTFE
Purchase
Properties
Related Categories Glass Stirrer Shafts, Glass Stirring Equipment, Glassware, Labware, Mixing Baffles More...
material   PTFE
mfr. no.   Ace Glass, 12177-148
shaft O.D. × L   10 mm × 330 mm
Description
General description
Ace-Thred thermometer/tubing adapters to mate glass or PTFE (stainless steel core) mixing baffles to standard taper joints. Baffles can be adjusted vertically or rotated to increase agitation.
Legal Information
Ace-Thred is a trademark of Ace Glass, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?