• USA Home
  • Z685089 - Ace sparger with hose barb
EMAIL THIS PAGE TO A FRIEND
Z685089 Aldrich
Ace sparger with hose barb
stopcock size 0-5 mm, L × O.D. 775 mm × 14 mm, porosity 70-100 μm
Purchase
Properties
Related Categories Dispersion tubes, Gas Tubes and Traps, Glassware, Labware
mfr. no.   Ace Glass, 6452-33
L × O.D.   775 mm × 14 mm
stopcock size   0-5 mm
pore size   70-100 μm porosity
Description
General description
Sparger tube with Ace fritted bottoms are available in various porosities. Top has either 0-3 or 0-5 mm stopcock and a 90 degree side arm hose connection. 10 mm O.D. spargers fit inside #11 Ace-Threds, 14 mm O.D. within #15 Ace- Threds.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?