• USA Home
  • Z685143 - Ace powder funnel with flat side
EMAIL THIS PAGE TO A FRIEND
Z685143 Aldrich
Ace powder funnel with flat side
top diam. 100 mm, Joint: ST/NS 45/50
Purchase
Properties
Related Categories Addition/Dropping, Funnels, Glass Funnels, Glassware, Labware,
material   borosilicate glass
Joint   Joint: ST/NS45/50
mfr. no.   Ace Glass
top diam.   100 mm
Description
General description
Ace powder funnel with standard taper joint and one flattened side for easy of use with multi-neck flasks.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?