• USA Home
  • Z685607 - Tygon® formula 2375 laboratory tubing

EMAIL THIS PAGE TO A FRIEND
Z685607 Aldrich

Tygon® formula 2375 laboratory tubing

I.D. × O.D. 2.4 mm × 4.0 mm, L 15 m (50 ft)

Purchase

Properties

Related Categories Labware, Tubing, Tygon Tubing More...
mfr. no.   St. Gobain, AJK00004
I.D. × O.D.   2.4 mm × 4.0 mm
L   15 m (50 ft)
wall thickness   0.79 mm

Description

General description

Tygon laboratory tubing, formula 2375, is chemically resistant tubing. Clear, flexible tubing suitable for specialty chemical production and hazardous material handling.

• Resistant to MEK, acids, bases, ketones, and alcohols
• Plasticizer free
• No contamination from leaching
• Biocompatible
• No disposal concerns
• Releases only carbon dioxide and water upon incineration
• Sterilize by gas or radiation

Specifications

Technical data:
Tensile Strength: 1900 psi (13.1 MPa)
Ultimate Elongation: 850%
Tensile Stress @ 100% elongation: 425 psi (2.9 MPa)
Tensile Set (@ 75% of ultimate elongation): 300%
Tear Resistance: 240 lb-f/inch (42 kN/m)
Compression Set Constant Deflection: 100% @ 158°F (70°C) for 22 hours
Brittle Temperature: < -75°F (<-59°C)
Low Temp. Flexibility: -103°F (-75°C)
Specific Gravity: 0.9
Water Absorption: 0.04% 24 hrs at 73°F (23°C)
Maximum Recommended Operating Temperature: 130°F
Color: Clear

Legal Information

Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?