• USA Home
  • Z740895 - Aldrich® Essentials graduated cylinder, class B, meets DIN ISO 4788
EMAIL THIS PAGE TO A FRIEND
Z740895 Aldrich
Aldrich® Essentials graduated cylinder, class B, meets DIN ISO 4788
capacity 10 mL, subdivision, 0.20 mL, 0.20 mL tol., Graduated "To Contain", plastic bumper, hex base
Purchase
Properties
Related Categories Cylinders, Graduated, Glassware, Labware, SIAL Essentials
feature   subdivision 0.20 mL
packaging   pkg of 6 ea
measuring range   0.20 mL tol.
capacity   10 mL
Description
General description
Clear borosilicate glass graduated cylinders with safety hexagonal base. Pour spout is offset of base point so that if cylinder tips the spout won’t be damaged. Hex base prevents the cylinder from rolling off a bench. Meets DIN ISO 4788 Class B tolerances and specifications. Complete with plastic bumper. Graduated “To Contain”.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?