• USA Home
  • Z740919 - Aldrich® Essentials graduated cylinder, class B, meets DIN ISO 4788
EMAIL THIS PAGE TO A FRIEND
Z740919 Aldrich
Aldrich® Essentials graduated cylinder, class B, meets DIN ISO 4788
capacity 1000 mL, subdivision, 10.0 mL, 10.0 mL tol., Graduated "To Contain", plastic bumper, hex base
Purchase
Properties
Related Categories Cylinders, Graduated, Glassware, Labware, SIAL Essentials
feature   subdivision 10.0 mL
packaging   pkg of 1 ea
measuring range   10.0 mL tol.
capacity   1000 mL
Description
General description
Clear borosilicate glass graduated cylinders with safety hexagonal base. Pour spout is offset of base point so that if cylinder tips the spout won’t be damaged. Hex base prevents the cylinder from rolling off a bench. Meets DIN ISO 4788 Class B tolerances and specifications. Complete with plastic bumper. Graduated “To Contain”.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?