• USA Home
  • Z740929 - Aldrich® Essentials volumetric pipette, class A, meets ASTM E 969
EMAIL THIS PAGE TO A FRIEND
Z740929 Aldrich
Aldrich® Essentials volumetric pipette, class A, meets ASTM E 969
capacity 100 mL, 0.080 mL tol., yellow
Purchase
Properties
Related Categories Glassware, Labware, Pipettes, Volumetric, SIAL Essentials
material   borosilicate glass
feature   class A
packaging   pkg of 12 ea
mfr. no.   Sigma-Aldrich
measuring range   0.080 mL tol.
capacity   100 mL
color   yellow
Description
General description
Borosilicate glass, reusable, volumetric pipets, Calibrated to contain. Permanent white fill mark and capacity mark. Color coded. Meets Class A, ASTM E 969 specifications.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?