• USA Home
  • Z740948 - Aldrich® Essentials culture tube with screw cap and PTFE/rubber liner
EMAIL THIS PAGE TO A FRIEND
Z740948 Aldrich
Aldrich® Essentials culture tube with screw cap and PTFE/rubber liner
capacity 30 mL, O.D. × L 25 mm × 100 mm, round bottom, round bottom
Purchase
Properties
Related Categories Glassware, Labware, SIAL Essentials, Tubes
packaging   pkg of 192 ea
  pkg of 192 ea
O.D. × L   25 mm × 100 mm
capacity   30 mL
Description
General description
Clear borosilicate glass, round bottom culture tubes with thread top and black phenolic cap. Cap has bonded PTFE faced rubber liner.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?