• USA Home
  • Z740957 - Aldrich® Essentials culture tube with screw cap and rubber liner
EMAIL THIS PAGE TO A FRIEND
Z740957 Aldrich
Aldrich® Essentials culture tube with screw cap and rubber liner
capacity 5 mL, O.D. × L 15 mm × 75 mm
Purchase
Properties
Related Categories Glassware, Labware, SIAL Essentials, Tubes
O.D. × L   15 mm × 75 mm
capacity   5 mL
Description
General description
Clear borosilicate glass, round bottom culture tubes with thread top and black phenolic cap. Cap has plain rubber liner. Conforms to ASTM E982.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?