• USA Home
  • Z740962 - Aldrich® Essentials culture tube with screw cap and rubber liner
EMAIL THIS PAGE TO A FRIEND
Z740962 Aldrich
Aldrich® Essentials culture tube with screw cap and rubber liner
capacity 15 mL, O.D. × L 15 mm × 150 mm
Purchase
Properties
Related Categories Glassware, Labware, SIAL Essentials, Tubes
O.D. × L   15 mm × 150 mm
capacity   15 mL
Description
General description
Clear borosilicate glass, round bottom culture tubes with thread top and black phenolic cap. Cap has plain rubber liner. Conforms to ASTM E982.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?