• USA Home
  • Z741118 - Q-Tube Pressure Monitor
EMAIL THIS PAGE TO A FRIEND
Z741118 Aldrich
Q-Tube Pressure Monitor
12 SS, pkg of 1 ea
Synonym: Q-Tube Pressure Monitor
Purchase
Properties
Related Categories Labware, Pressure Vessels, Q-Tube Pressure Tube Reactors More...
material   316 stainless steel adapter (with matched PTFE sleeve)
packaging   pkg of 1 ea
mfr. no.   Q Labtech, 100-PRMSET-12-SS
Description
General description
The parts with direct chemical contact are made of 316 stainless steel, PTFE, Simriz, or glass.
Q-Tube Pressure Monitor-12-SS contains all items needed for immediate use:
• 316 stainless steel Adapter and matched PTFE sleeve
• 1 pressure gauge-300 psi
• 25 sets of pressure tube-12 mL/septa-12, etc.
Legal Information
Q-Tube is a trademark of Q Labtech LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?