• USA Home
  • Z741639 - Whatman® indicator papers

EMAIL THIS PAGE TO A FRIEND
Z741639 Aldrich

Whatman® indicator papers

Specialized Test Papers, Lead Acetate

Synonym: Whatman filter, filter papers, paper filter

Purchase

Properties

feature   Specialized Test Papers, Lead Acetate
packaging   pkg of 1 ea
mfr. no.   Whatman Article No., 28418899 (US reference)
  Whatman, 2602-501A
W × L   7 mm × 50 mm

Description

Features and Benefits

• Instant pH readings
• Accurate for a wide range of routine pH testing
• Inexpensive
• Convenient and portable for field use
• pH Indicators

General description

Lead Acetate Test Paper
Used for detecting hydrogen sulfide, this rapid qualitative test paper when wetted with distilled water can detect as little as 5 ppm of H2S in the atmosphere or in a gas stream. Hydrogen peroxide can be detected with this paper by preblackening the paper in H2S. Concentrations as low as 4 ppm can be detected.

Whatman offers a range of pH indicator and test papers to meet your specific needs. Made with traditional Whatman quality, these products combine ease-of-use with unsurpassed accuracy and consistency.

The convenience of using indicator papers for the rapid determination of pH values has led to many applications in laboratories and industry.

Other Notes

Global SIAL customers (excluding Japan) can now order any product from the GE Healthcare Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.

The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of GE Healthcare

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

WHA2602501A Whatman® indicator papers, Specialized Test Papers, Lead Acetate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?