• USA Home
  • Z755354 - 3D Biotek 3D perfusion bioreactor system with pump

EMAIL THIS PAGE TO A FRIEND
Z755354 Aldrich

3D Biotek 3D perfusion bioreactor system with pump

24 well chamber with PS scaffold inserts, autoclavable, AC input 120 V

Purchase

Properties

Related Categories 3D Bioreactors, 3D Biotek Bioreactor System, 3D Cell Culture Products, Cell Culture Supplies, Labware More...
material   polycarbonate (4 interchangeable chambers)
sterility   non-sterile
feature   CSA approved
  autoclavable
  single use bioreactor system
mfr. no.   3D Biotek
W × D × H   14 in. × 13 in. × 13 in.
matrix active group   non-treated surface treatment
suitability   suitable for (stem cell growth and differentiation)

Description

Application

Numerous studies have proven that cells grow better under dynamic culture condition, simply because both nutrition supply and metabolic waste removal are better under dynamic culture conditions. Additionally, in some instances, the shear force produced by the flowing medium can act as a mechanical stimuli signal that will further promote stem cell differentiation toward certain cell lineage, such as osteoblast. Combined with the 3D Insert scaffold technology, the 3D Perfusion Bioreactor can facilitate the promotion of stem cell growth and differentiation, thereby enhancing stem cell research.

General description

The 3D perfusion bioreactor consists of multiple independent, autoclavable polycarbonate chambers. The chambers are interchangeable and are specially designed to take advantage of 3D Insert scaffolds with various sizes ranging from 96-well to 6-well. Cell culture medias are perfused through the open porous structure of scaffolds using a pulsatile pump. The entire unit is autoclavable (except pump, of course) and can be used as a single-use bioreactor system.

Legal Information

3D Insert is a trademark of 3D Biotek, LLC

Price and Availability

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?