• USA Home
  • D-090 - 3-Desmethylprodine hydrochloride solution

EMAIL THIS PAGE TO A FRIEND
D-090 Cerilliant

3-Desmethylprodine hydrochloride solution

1.0 mg/mL in acetonitrile (as free base), ampule of 1 mL, certified reference material

  • CAS Number 4968-48-3

  • Empirical Formula (Hill Notation) C15H21NO2 · HCl

  • Molecular Weight 283.79

Purchase

Properties

Related Categories Additional Standards, Analgesics (non-opiates), Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs),
grade   certified reference material
InChI Key   WQANBUZBKNCRKH-UHFFFAOYSA-N
feature   (Snap-N-Spike®)
packaging   ampule of 1 mL
drug control   Narcotic Licence Schedule A (Switzerland)
concentration   1.0 mg/mL in acetonitrile (as free base)
format   single component solution
storage temp.   −20°C

Description

General description

A certified Snap-N-Spike® Solution suitable for LC/MS or GC/MS methods and applications in clinical toxicology, forensic analysis, urine drug testing, or pain prescription monitoring. MPPP or 3-Desmethylprodine is an opioid analgesic developed in the 1940s for use as a pharmaceutical but abused illegally today as a recreational drug. This designer drug is an analog of meperidine (Demerol®) but is listed by US DEA as a Schedule I drug since it has no medicinal use.

Legal Information

Demerol is a registered trademark of Sanofi-Aventis

Snap-N-Spike is a registered trademark of Cerilliant Corporation

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

Emerging Drugs of Abuse

The illicit drug market is dynamic with new drugs of abuse constantly appearing. In addition to synthetic cannabinoids and synthetic cathinones, recent emerging drugs of abuse include new analogs of ...
Keywords: Addition reactions

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

M-036 Meperidine-D4 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material
P-071 Pain Management Multi-component Opiate Mixture-13 solution, 100 μg/mL each component (10 μg/mL Fentanyl), ampule of 1.0 mL, certified reference material
O-020 Opiate Multi-component Mixture-5 solution, 250 μg/mL each component in methanol, ampule of 1.0 mL, certified reference material
M-035 Meperidine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
M-038 Meperidine-D4 solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
N-017 Normeperidine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material
N-020 Normeperidine-D4 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material
N-083 Normeperidine-D4 solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
94791 SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules
43917 SnapIT Ampule Opener, large, for 5-10ml, 10-15ml, 20-25ml ampoules

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?