Synonym:
1,3,3-
CAS Number 470-82-6
Empirical Formula (Hill Notation) C10H18O
Molecular Weight 154.25
Beilstein Registry Number 105109
EC Number 207-431-5
MDL number MFCD00167977
| Related Categories | A - D, Achillea, Alphabetical, Analytical Standards, Analytical/Chromatography, |
| grade | primary reference standard |
| refractive index |
n |
| bp | 176-177 °C(lit.) |
| mp | 1-2 °C(lit.) |
| density | 0.921 g/mL at 25 °C(lit.) |
| storage temp. | 2-8°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?