00185 Fluka

Acetamide Nutrient Broth

for microbiology

DOWNLOAD MSDS (PDF)

Properties

Related Categories A - M, All Liquid Media/Broths, Biochemical Identification Media, A to Z, Identification Tests & Reagents, Media,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
composition   acetamide, 2.0 g/L
  dipotassium hydrogen phosphate, 0.2 g/L
  ferrous sulfate, 0.0005 g/L
  magnesium sulfate, 0.158 g/L
  sodium chloride, 0.2 g/L
  sodium molybdate, 0.005 g/L
final pH   7.0±0.2 (25 °C)
suitability   selective by defined nutrients for Pseudomonas

Description

Application

Synthetic medium to detect microbial utilization of acetamide. Corresponds to the suggestions of the "Deutsches Institut für Normung (DIN)" and the test procedures set out in §35 LMBG for the examination of water for Pseudomonas aeruginosa.

Other Notes

Decomposition of acetamide as taxonomic marker for some species of the genus Pseudomonas1; Comparative studies on the enrichment of P. aeruginosa2

Price and Availability

Customers Also Viewed

Acetamide
695122Acetamide

sublimed, 99%

Acetamide

~99% (GC)

for microbiology

for microbiology

for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?