Synonym:
2-
CAS Number 6151-25-3
Empirical Formula (Hill Notation) C15H10O7 · 2H2O
Molecular Weight 338.27
Beilstein Registry Number 317313
Colour Index Number 75670
EC Number 204-187-1
MDL number MFCD00149487
| Related Categories | Aesculus, Allium, Alphabetical, Analytical Standards, Analytical/Chromatography, |
| grade | primary reference standard |
| CofA | certificate is enclosed in each package |
| mp | >300 °C(lit.) |
Reference standard in the analysis of herbal medicinal products.
Produced and qualified by HWI Analytik
Exact content by quantitative NMR can be found on the certificate
Quercetin dihydrate is a flavonoid with anticancer activity. Quercetin is a mitochondrial ATPase and phosphodiesterase inhibitor. It Inhibits PI3-kinase activity and slightly inhibits PIP kinase activity. Quercetin has antiproliferative effects on cancer cell lines, reduces cancer cell growth via type II estrogen receptors, and arrests human leukemic T cells in late G1 phase of the cell cycle.
Customers Also Viewed
≥95% (HPLC), solid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?