CAS Number 9004-54-0
Linear Formula [C6H10O5]n
EC Number 232-677-5
MDL number MFCD00130935
| Related Categories | Analytical Standards, Analytical/Chromatography, Application CRMs, Biochemicals and Reagents, Carbohydrates, |
| grade | analytical standard |
| for GPC | |
| CofA | exact values can be taken from the accompanying certificate of analysis |
| mol wt | Mn ~8,110 |
| Mp ~9,890 | |
| Mw ~11,600 | |
| Mw/Mn | ~1.43 |
TSKgel® HPLC columns for Gel Permeation Chromatography are available in eight pore sizes, ranging from an exclusion limit of ~2,000 Daltons for G1000HHR columns to more than 10,000,000 Daltons for
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?